Learn More
Trifluralin, TRC
Product Code. 30138118
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138118 1 g 1g
17919389 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138118 Supplier Toronto Research Chemicals Supplier No. TRCT7935501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1582-09-8
Molecular Formula C13 H16 F3 N3 O4
Molecular Weight (g/mol) 335.28
Synonym p-Toluidine, α,α,α-trifluoro-2,6-dinitro-N,N-dipropyl- (6CI,8CI), 2,6-Dinitro-N,N-dipropyl-4-(trifluoromethyl)benzenamine, 2,6-Dinitro-4-(trifluoromethyl)-N,N-dipropylaniline, 2,6-Dinitro-N,N-di-n-propyl-α,α,α-trifluoro-p-toluidine, 2,6-Dinitro-N,N-dipropyl-4-(trifluoromethyl)aniline, 4-(Di-n-propylamino)-3,5-dinitro-1-(trifluoromethyl)benzene, 4-(Trifluoromethyl)-2,6-dinitro-N,N-dipropylaniline, Agriflan, Agriflan 24, Bayonet, Biobarrier, Brassix, Elancolan, L 36352, Lilly 36,352, N,N-Di-n-propyl-2,6-dinitro-4-trifluoromethylaniline, N,N-Dipropyl-2,6-dinitro-4-(trifluoromethyl)aniline, N,N-Dipropyl-4-trifluoromethyl-2,6-dinitroaniline, Nitran, Nitran K, Olitref, Premerlin, Super-Treflan, Synfloran, Tefralin, Trefanocide, Treflan, Treflan 4E, Trifloran, Trifluralin, Trifluraline, Triflurex, Triflurom, Triflurotox, Trifsan, Trikepin, Trilan, Trilan (fungicide), Trilin, Tristar, Zeltoxone, α,α,α-Trifluoro-2,6-dinitro-N,N-di-n-propyl-p-toluidine, α,α,α-Trifluoro-2,6-dinitro-N,N-dipropyl-p-toluidine
IUPAC Name 2,6-dinitro-N,N-dipropyl-4-(trifluoromethyl)aniline
SMILES CCCN(CCC)c1c(cc(cc1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C13H16F3N3O4/c1-3-5-17(6-4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3
Formula Weight 335.1093
Chemical Name or Material Trifluralin