accessibility menu, dialog, popup

UserName

CAS RN 1582-09-8

H3CNCH3NOOFFFNOO

CAS RN 1582-09-8

IUPAC Name: 2,6-dinitro-N,N-dipropyl-4-(trifluoromethyl)aniline
Synonyms: tristar trifluralin (iso) trifluralin
Molecular Weight (g/mol): 335.28
Molecular Formula: C13H16F3N3O4
InChi Key: ZSDSQXJSNMTJDA-UHFFFAOYSA-N
SMILES: CCCN(CCC)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (7)
Solvent or Matrix 
  • (2)
  • (2)
  • (3)
Concentration 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (7)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)