Learn More
Tri(nonylphenyl) Phosphite, TRC
Product Code. 30010765
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010765 100 mg 100mg
30007233 1 g 1g
30004191 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010765 Supplier Toronto Research Chemicals Supplier No. TRCP049950100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26523-78-4
Molecular Formula C45 H69 O3 P
Molecular Weight (g/mol) 689
Synonym Tri(monononylphenyl) phosphite, Weston TNPP, Weston 399, Wytox 312, Alkanox 70, Antage TNP, Antioxidant 1178, Antioxidant TNPP, Crystol EPR 3400, Doverphos 4, Doverphos HP 4HR, Doverphos HiPure 4HR, Doverphos Highpure 4, Irgafos TNPP, Mark 1178, Naugard TNPP, Nonylphenyl phosphite, Naugard TNPP, Nonylphenyl phosphite, Sumilizer TNP
IUPAC Name Tri(nonylphenyl) Phosphite
SMILES CCCCCCCCCc1ccccc1OP(Oc2ccccc2CCCCCCCCC)Oc3ccccc3CCCCCCCCC
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C45H69O3P/c1-4-7-10-13-16-19-22-31-40-34-25-28-37-43(40)46-49(47-44-38-29-26-35-41(44)32-23-20-17-14-11-8-5-2)48-45-39-30-27-36-42(45)33-24-21-18-15-12-9-6-3/h25-30,34-39H,4-24,31-33H2,1-3H3
Formula Weight 688.4984
Chemical Name or Material Tri(nonylphenyl) Phosphite