accessibility menu, dialog, popup

UserName

CAS RN 26523-78-4

H3COPOCH3OCH3

CAS RN 26523-78-4

IUPAC Name: tris(4-nonylphenyl) phosphite
Synonyms: tris(nonylphenyl)phosphite
Molecular Weight (g/mol): 689.02
Molecular Formula: C45H69O3P
InChi Key: MGMXGCZJYUCMGY-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(OP(OC2=CC=C(CCCCCCCCC)C=C2)OC2=CC=C(CCCCCCCCC)C=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results