missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TREM2 Monoclonal specifically detects TREM2 in Mouse samples. It is validated for ELISA/Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | TREM2 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | 14 |
| Conjugate | DyLight 755 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TREM2 (NP_112544.1; Met1-Pro168). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?