Learn More
trans-3-Aminocyclohexanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
Product Code. 15447186
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15447186
Brand: Thermo Scientific Alfa Aesar H62232.03
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water, glycerol. Insoluble in ethanol, chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 862401-49-8
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD11040475
InChI Key BOUHXQPJYJANEO-KGZKBUQUSA-N
Synonym trans-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hcl, 1r,3r-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hydrochloride, trans-3-aminocyclohexane carboxylic acid hydrochloride, trans-3-aminocyclohexane-1-carboxylic acid hydrochloride
PubChem CID 46172878
IUPAC Name (1R,3R)-3-aminocyclohexane-1-carboxylic acid;hydrochloride
SMILES C1CC(CC(C1)N)C(=O)O.Cl
Specifications
Solubility Information Soluble in water,glycerol. Insoluble in ethanol,chloroform
Quantity 1 g
Formula Weight 179.64
Percent Purity 97%
Chemical Name or Material trans-3-Aminocyclohexanecarboxylic acid hydrochloride
compliance-icons
Product Identifier
  • trans-3-Aminocyclohexanecarboxylic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : trans-3-Aminocyclohexanecarboxylic acidhydrochloride