accessibility menu, dialog, popup

UserName

CAS RN 862401-49-8

NH2OHOHCl

CAS RN 862401-49-8

IUPAC Name: 3-aminocyclohexane-1-carboxylic acid hydrochloride
Synonyms: 3-aminocyclohexane-1-carboxylic acid hydrochloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: BOUHXQPJYJANEO-UHFFFAOYNA-N
SMILES: Cl.NC1CCCC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)