missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAM 39
Description
TRAM 39 is a potent intermediate conductance Ca2+-activated K+ channel (KCa3.1) blocker (Kd = 60 nM). Has no effect on cytochrome p450 activity. Inhibits I-EBIO -stimulated increases in rat artery membrane potential ex vivo. Also diminishes LPS-induced cryptidin (mammalian α-defensin) release from paneth cells in vitro.
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-α,α-diphenylbenzeneacetonitrile |
| CAS | 197525-99-8 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Molecular Formula | C20H14ClN |
| Formula Weight | Observed MW: 303.78 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?