Learn More
Invitrogen™ TR-FRET Dilution Buffer
Product Code. 10398323
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10398323 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10398323 Supplier Invitrogen™ Supplier No. PV3574
 Restricted Item
Estimated Shipment: 26-11-2026
TR-FRET Dilution Buffer
TR-FRET Dilution Buffer is an essential component for kinase assays using LanthaScreen™ technology. The TR-FRET Dilution Buffer is used to dilute LanthaScreen™ antibodies for screening applications.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dilution Buffer
Physical Form Liquid
Quantity 100 mL