Learn More
Invitrogen™ TR-FRET Coregulator Buffer E
Product Code. 10524614
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10524614 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10524614 Supplier Invitrogen™ Supplier No. PV4540
 Restricted Item
Estimated Shipment: 02-12-2026
TR-FRET Coregulator Buffer E
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TR-FRET Coregulator Buffer
Physical Form Liquid
Quantity 25 mL