missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tocriscreen FDA-Approved Drugs Library

Description
Tocriscreen FDA-Approved Drugs Library is a library of 190 compounds equivalent to the pharmacologically active components of drug formulations approved by the FDA. Compounds supplied pre-dissolved in DMSO (100 μL, 10 mM solution). Contains compounds covering a diverse range of pharmaceutical targets and therapeutic areas e.g. immunology, oncology. Includes both established therapeutics, e.g. tamoxifen (Cat. No. 0999) & paclitaxel (Cat. No. 1097), as well as more recently approved drugs such as darunavir (Cat. No. 6710) & sorafenib (Cat. No. 6814). Ideal for in vitro and in vivo drug repurposing/repositioning studies.
This library replaces the Tocriscreen Library of FDA-Approved Compounds (Cat. No. 5932).
All Tocris products are for biomedical research use only. They are not intended for human or veterinary use.
For more information about this compound library, please download our informative flyer .
Specifications
Specifications
| Quantity | 1 Set |
| Target | Screening Libraries |
| Formula Weight | 22978 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?