missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTpro 18-plex, TMTpro-134C, and TMTpro-135N Label Reagents

Product Code. 17901861 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mg
4 x 50 μg (per tag)
6 x 0.5 mg
Label or Dye:
TMTpro 18-plex
TMTpro-134C and TMTpro-135N
Unit Size:
3mg
5mg
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Label or Dye Unit Size
17901861 4 x 50 μg (per tag) TMTpro 18-plex Each
17496572 1 x 5 mg TMTpro 18-plex 5mg
17331783 1 x 5 mg TMTpro-134C and TMTpro-135N 5mg
17418273 6 x 0.5 mg TMTpro 18-plex 3mg
17423772 6 x 0.5 mg TMTpro-134C and TMTpro-135N 3mg
17483812 6 x 0.5 mg TMTpro-134C and TMTpro-135N 3mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17901861 Supplier Thermo Scientific™ Supplier No. A58335
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TMTpro 18plex Label Reagents enable multiplexing of up to 18 samples. Addition of TMTpro 134C and TMTpro 135N Label Reagents increase sample multiplexing from 16-plex to 18-plex, enabling even greater throughput for protein identification and quantitation using tandem mass spectrometry (MS).

TMTpro label reagents are the next generation of tandem mass tags, designed to increase the level of sample multiplexing without compromising on protein identification and quantitation. The Thermo Scientific TMTpro 18plex Label Reagent Set enables multiplexing up to 18 samples, enabling even greater throughput for protein identification and quantitative analysis by tandem mass spectrometry (MS).

The structures of the TMTpro and TMT tags are similar in being isobaric and amine reactive, but the TMTpro tag has a longer spacer region and isobutyl proline mass reporter. After MS/MS fragmentation, each TMTpro 18plex tag generates a unique reporter mass (e.g., TMTpro 126-135 Da) in the low-mass region of the high-resolution MS/MS spectrum that is used for relative quantitation of protein expression levels.

The TMT pro134C and TMTpro135N Label Reagents are identical in composition and structure to the other TMTpro 16plex tags, but are not isobaric. Addition of the TMTpro 134C and 135N tags create a 17th and 18th channel for relative quantitation using high resolution Orbitrap MS instruments. These tags are available separately (Cat. No. A52046, A52048), as part of the TMTpro 18plex Label Reagent Set, or as part of the complete 18plex set of reagents in a 96-well plate format.

Features of the TMTpro 18plex Label Reagent Set include:

  • Multiplex—concurrent MS analysis of up to 18 samples derived from cells, tissues, or biological fluids
  • Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
  • Efficient—amine-reactive, NHS ester-activated reagents ensure greater than 95% labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
  • Optimized for use—with high resolution Thermo Scientific MS/MS Orbitrap platforms, analysis fully supported by Proteome Discoverer 2.3
  • Convenient size—provided in a ready-to-use, single-use format to ensure optimal stability and performance

For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES902 or ES903). When combined with the industry-leading, high resolution Orbitrap instruments and software, TMTpro reagents provide an integrated total solution for quantitative protein expression analysis.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Liquid in stabilized DMSO solution
For Use With (Equipment) Mass Spectrometer
Product Type Isobaric Label Reagent Set
Product Line TMT
Sufficient For 72 Reactions
Content And Storage

Sufficient for: Four 18-plex (4 x 18-way) experiments
• TMTpro-126 Label Reagent, 4 x 50 μg
• TMTpro-127 N Label Reagent, 4 x 50 μg
• TMTpro-127 C Label Reagent, 4 x 50 μg
• TMTpro-128 N Label Reagent, 4 x 50 μg
• TMTpro-128 C Label Reagent, 4 x 50 μg
• TMTpro-129 N Label Reagent, 4 x 50 μg
• TMTpro-129 C Label Reagent, 4 x 50 μg
• TMTpro-130 N Label Reagent, 4 x 50 μg
• TMTpro-130 C Label Reagent, 4 x 50 μg
• TMTpro-131 N Label Reagent, 4 x 50 μg
• TMTpro-131 C Label Reagent, 4 x 50 μg
• TMTpro-132 N Label Reagent, 4 x 50 μg
• TMTpro-132 C Label Reagent, 4 x 50 μg
• TMTpro-133 N Label Reagent, 4 x 50 μg
• TMTpro-133 C Label Reagent, 4 x 50 μg
• TMTpro-134 N Label Reagent, 4 x 50 μg
• TMTpro-134 C Label Reagent, 4 x 50 μg
• TMTpro-135 N Label Reagent, 4 x 50 μg

Store at -20°C.

Label or Dye TMTpro 18-plex
Starting Material Cell Lysate
Final Product Type Protein PEPTIDE
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 4 x 50 μg (per tag)
Form Liquid
Show More Show Less
How can I tell if the buffer in which my peptides are resuspended are compatible with TMT and TMTpro labels?

TMT and TMTpro labels are not compatible with buffers containing primary amines. After protein digestion, peptides need to be cleaned up and dried down. Prior to labeling, we do recommend resuspending the peptides in 10 µL of either 100 mM TEAB (pH 8.5) or 100 mM HEPES (pH 8.0).

Is the TMT stabilizing solution compatible with subsequent MS steps?

For further steps after TMT labelling, the stabilizing solution can be considered as a 33% DMSO solution. If subsequent excess TMT removal will be performed using EasyPep solid-phase extraction (SPE), the DMSO solution does not need to be diluted. On the other hand, the use of any reverse phase chromatography method (i.e., Cat. No. 84868) might need dilution of the sample to ensure that the DMSO concentration is tolerated by the resin. Alternatively, the peptide mix can be dried down using Speed Vac and then resuspended in the corresponding buffer for a particular column (Reverse Phase or Desalting).

With TMT 10plex, TMT 11plex, TMTpro 16plex, and TMTpro 18plex 96-well plates, how much protein can be used per TMT or TMTpro reaction?

TMT and TMTpro label reagents in 96-well plates are optimized to work with 1 to 10 µg of protein digest.

Can the sealing foil be partially removed with TMT 10plex, TMT 11plex, TMTpro 16plex, and TMTpro 18plex 96-well plates?

Yes, the sealing foil can be partially peeled to access individual sets of TMT or TMTpro labels. Alternatively, each of the wells can be perforated to transfer the TMT/TMTpro labels to a different tube, if needed.

How do I determine the correct orientation of the TMT 10plex, TMT 11plex, TMTpro 16plex, and TMTpro 18plex 96-well plates?

All the plates are barcoded in the long lower edge, helping to recognize correct plate orientation, plus the plate has a notch on the left corners.

Would the TMT and TMTpro labels be affected if the TMT 10plex, TMT 11plex, TMTpro 16plex, and TMTpro 18plex 96-well plates are going through several freeze-thaw cycles?

The TMT and TMTpro labels from TMT 10plex, TMT 11plex, TMTpro 16plex and TMTpro 18plex 96-well plates are stable after 9 freeze-thaw cycles, making it possible to use each of the experiment sets at different times.

With TMT 10plex, TMT 11plex, TMTpro 16plex, and TMTpro 18plex 96-well plates, can I run each of the experiment sets in different runs or should I use the plate all at once?

The entire plate can be run at one time or each experiment set can be run at a different time, using a high-throughput automation platform.

How many sets of experiments can I run with TMTpro 18plex Label Reagents ​in stabilized solution, ​96-well plate, 4 x 50 ug​ (Cat. No. A58335)?

TMTpro 18plex 96-well plate (Cat. No. A58335) contains 4 independent sets of experiments, each of them occupying three columns with rows A and H empty. Please see the user guide for further plate layout details.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.