missing translation for 'onlineSavingsMsg'
Learn More

TIMM13 Antibody, Novus Biologicals™

Product Code. 18215001 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18215001 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215001 Supplier Novus Biologicals Supplier No. NBP174226
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIMM13 Polyclonal specifically detects TIMM13 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P62075
Gene Alias mitochondrial import inner membrane translocase subunit Tim13, mitochondrial import inner membrane translocase subunit Tim13B, ppv1, Tim13, TIM13B, TIMM13A, TIMM13B, translocase of inner mitochondrial membrane 13 (yeast) homolog B, translocase of inner mitochondrial membrane 13 homolog (yeast)
Gene Symbols TIMM13
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the N terminal of Timm13. Immunizing peptide sequence FGSDFGGTGGGKLDPGAIMEQVKVQIAVANAQELLQRMTDKCFRKCIGKP.
Molecular Weight of Antigen 10 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26517
Test Specificity This product is specific to Subunit or Isoform: Tim13.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.