missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | TIM-3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 108 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human TIM-3 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?