missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thioredoxin-2 Monoclonal specifically detects Thioredoxin-2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Thioredoxin-2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 56 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | mitochondrial thioredoxin, MTRX, MT-TRX, thioredoxin 2, thioredoxin, mitochondrial, thioredoxin-2, TRX2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human Thioredoxin-2 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?