Learn More
Thiophene-2-carboxylic acid, 99%
Product Code. 11591696
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11591696 100 g Each
11464164 25 g 25g
11474164 100 g 100g
11484164 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11591696 Supplier Amresco Supplier No. M252100ML
CAS: 527-72-0 | C5H4O2S | 128.15 g/mol
A hypoglycemic, anti-lipolytic agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS 527-72-0
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.15
MDL Number MFCD00005437
InChI Key QERYCTSHXKAMIS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
PubChem CID 10700
ChEBI CHEBI:71241
IUPAC Name thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=CS1
Specifications
Melting Point 126°C to 130°C
Density 1.42
Boiling Point 259°C to 261°C
Quantity 100 g
Beilstein 110150
Merck Index 14,9354
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material Thiophene-2-carboxylic acid