accessibility menu, dialog, popup

UserName

CAS RN 527-72-0

IUPAC Name: thiophene-2-carboxylic acid
Synonyms: thiophene-2-carboxylic acid
Molecular Weight (g/mol): 128.15
Molecular Formula: C5H4O2S
InChi Key: QERYCTSHXKAMIS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CS1
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)