missing translation for 'onlineSavingsMsg'
Learn More

Thiometon-sulfone, TRC

Product Code. 30134984
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134984 250 mg 250mg
18021340 25 mg 25mg
18071830 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134984 Supplier Toronto Research Chemicals Supplier No. TRCT344670250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20301-63-7
Molecular Formula C6 H15 O4 P S3
Molecular Weight (g/mol) 278.35
Synonym Thiometon-sulfone, Ethanethiol, 2-(ethylsulfonyl)-, S-ester with O,O-dimethyl phosphorodithioate (8CI), Thiometon sulfone
IUPAC Name 2-ethylsulfonylethylsulfanyl-dimethoxy-sulfanylidene-λ^{5}-phosphane
SMILES CCS(=O)(=O)CCSP(=S)(OC)OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H15O4PS3/c1-4-14(7,8)6-5-13-11(12,9-2)10-3/h4-6H2,1-3H3
Formula Weight 277.987
Percent Purity >95%
Chemical Name or Material Thiometon-sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.