CAS RN 20301-63-7
CAS RN 20301-63-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thiometon-sulfone, TRC
CAS: 20301-63-7 Molecular Formula: C6 H15 O4 P S3 Molecular Weight (g/mol): 278.35 Synonym: Thiometon-sulfone,Ethanethiol, 2-(ethylsulfonyl)-, S-ester with O,O-dimethyl phosphorodithioate (8CI),Thiometon sulfone IUPAC Name: 2-ethylsulfonylethylsulfanyl-dimethoxy-sulfanylidene-λ^{5}-phosphane SMILES: CCS(=O)(=O)CCSP(=S)(OC)OC