missing translation for 'onlineSavingsMsg'
Learn More

Tetramisole hydrochloride, 99.6%, MP Biomedicals™

Product Code. 11458251 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458251 5 g 5g
11468251 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458251 Supplier MP Biomedicals Supplier No. 0215211905

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 83-67-0
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.167
InChI Key YAPQBXQYLJRXSA-UHFFFAOYSA-N
Synonym theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
PubChem CID 5429
ChEBI CHEBI:28946
IUPAC Name 3,7-dimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)NC(=O)N2C

Specifications

Assay Percent Range 0.996
Quantity 5 g
Formula Weight 240.8
Physical Form Powder
Chemical Name or Material Tetramisole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.