accessibility menu, dialog, popup

UserName

H3CNNNCH3ONHO

CAS RN 83-67-0

IUPAC Name: 3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: theobromine 可可碱
Molecular Weight (g/mol): 180.17
Molecular Formula: C7H8N4O2
InChi Key: YAPQBXQYLJRXSA-UHFFFAOYSA-N
SMILES: CN1C=NC2=C1C(=O)NC(=O)N2C
Molecular Weight (g/mol) 
  • (7)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (6)
  • (1)
Physical Form 
  • (3)
  • (5)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (7)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (6)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)