missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylmethoxydimethylsilane, TRC

Product Code. 30179197
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179197 5 g 5g
30179125 10 g 10g
30179162 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179197 Supplier Toronto Research Chemicals Supplier No. TRCB7000055G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66548-21-8
Molecular Formula C7H18OSi
Molecular Weight (g/mol) 146.3
Synonym (tert-Butyl)dimethyl(methoxy)silane, (1,1-Dimethylethyl)methoxydimethylsilane;
IUPAC Name tert-butyl-methoxy-dimethylsilane
SMILES C[Si](OC)(C(C)(C)C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H18OSi/c1-7(2,3)9(5,6)8-4/h1-6H3
Formula Weight 146.1127
Chemical Name or Material tert-Butylmethoxydimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.