CAS RN 66548-21-8
CAS RN 66548-21-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
tert-Butylmethoxydimethylsilane, TRC
CAS: 66548-21-8 Molecular Formula: C7H18OSi Molecular Weight (g/mol): 146.3 Synonym: (tert-Butyl)dimethyl(methoxy)silane,(1,1-Dimethylethyl)methoxydimethylsilane; IUPAC Name: tert-butyl-methoxy-dimethylsilane SMILES: C[Si](OC)(C(C)(C)C)C