missing translation for 'onlineSavingsMsg'
Learn More

TeNT Light Chain/Tetanus Toxin Antibody (TetE3), Novus Biologicals™

Product Code. 18214600 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214600 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214600 Supplier Novus Biologicals Supplier No. NBP105447
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TeNT Light Chain/Tetanus Toxin Monoclonal specifically detects TeNT Light Chain/Tetanus Toxin in Bacteria samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TeNT Light Chain/Tetanus Toxin
Applications Western Blot, ELISA
Classification Monoclonal
Clone TetE3
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Gene Alias Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain
Gene Symbols TETX
Host Species Mouse
Immunogen have toxin neutralization activity (in vivo assay)
Purification Method Protein G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1061100
Test Specificity have toxin neutralization activity (in vivo assay)
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.