Learn More
Tebufenpyrad, TRC
Product Code. 30135459
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135459 10 g 10g
18004030 1 g 1g
18031920 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135459 Supplier Toronto Research Chemicals Supplier No. TRCT01350010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119168-77-3
Molecular Formula C18 H24 Cl N3 O
Molecular Weight (g/mol) 333.86
Synonym 4-Chloro-N-[[4-(1,1-dimethylethyl)phenyl]methyl]-3-ethyl-1-methyl-1H-pyrazole-5-carboxamide (ACI), AC 801757, Fenpyrad, Masai, MK 239, N-(4-tert-Butylbenzyl)-4-chloro-3-ethyl-1-methylpyrazole-5-carboxamide, Oscar, Pyranica, Tebufenpyrad, 1H-Pyrazole-5-carboxamide, 4-chloro-N-[[4-(1,1-dimethylethyl)phenyl]methyl]-3-ethyl-1-methyl- (9CI, ACI)
IUPAC Name N-[(4-tert-butylphenyl)methyl]-4-chloro-5-ethyl-2-methylpyrazole-3-carboxamide
SMILES CCc1nn(C)c(C(=O)NCc2ccc(cc2)C(C)(C)C)c1Cl
Specifications
Quantity 10 g
InChI Formula InChI=1S/C18H24ClN3O/c1-6-14-15(19)16(22(5)21-14)17(23)20-11-12-7-9-13(10-8-12)18(2,3)4/h7-10H,6,11H2,1-5H3,(H,20,23)
Formula Weight 333.1608
Chemical Name or Material Tebufenpyrad