accessibility menu, dialog, popup

UserName

CAS RN 119168-77-3

CH3NNCH3OHNH3CCH3CH3Cl

CAS RN 119168-77-3

IUPAC Name: N-[(4-tert-butylphenyl)methyl]-4-chloro-3-ethyl-1-methyl-1H-pyrazole-5-carboxamide
Synonyms: tebufenpyrad pyranica C18H24clN3O
Molecular Weight (g/mol): 333.86
Molecular Formula: C18H24ClN3O
InChi Key: ZZYSLNWGKKDOML-UHFFFAOYSA-N
SMILES: CCC1=NN(C)C(C(=O)NCC2=CC=C(C=C2)C(C)(C)C)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
Element 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results