Learn More
Taspine, TRC
Product Code. 30138321
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138321 50 mg 50mg
30005874 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138321 Supplier Toronto Research Chemicals Supplier No. TRCT00780050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 602-07-3
Molecular Formula C20 H19 N O6
Molecular Weight (g/mol) 369.37
SMILES COc1ccc2C(=O)Oc3c(OC)cc(CCN(C)C)c4C(=O)Oc1c2c34
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C20H19NO6/c1-21(2)8-7-10-9-13(25-4)18-16-14(10)20(23)27-17-12(24-3)6-5-11(15(16)17)19(22)26-18/h5-6,9H,7-8H2,1-4H3
Formula Weight 369.1212
Percent Purity >95%
Chemical Name or Material Taspine
Grade HPLC