CAS RN 602-07-3
CAS RN 602-07-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Taspine, TRC
CAS: 602-07-3 Molecular Formula: C20 H19 N O6 Molecular Weight (g/mol): 369.37 SMILES: COc1ccc2C(=O)Oc3c(OC)cc(CCN(C)C)c4C(=O)Oc1c2c34