missing translation for 'onlineSavingsMsg'
Learn More

Tandospirone, TRC

Product Code. 30415131
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415131 5 mg 5mg
30420943 25 mg 25mg
30417334 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415131 Supplier Toronto Research Chemicals Supplier No. TRCT0064305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87760-53-0
Molecular Formula C21 H29 N5 O2
Molecular Weight (g/mol) 383.49
SMILES O=C1[C@@H]2[C@H]3CC[C@H](C3)[C@@H]2C(=O)N1CCCCN4CCN(CC4)c5ncccn5

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H29N5O2/c27-19-17-15-4-5-16(14-15)18(17)20(28)26(19)9-2-1-8-24-10-12-25(13-11-24)21-22-6-3-7-23-21/h3,6-7,15-18H,1-2,4-5,8-14H2/t15-,16+,17+,18-
Formula Weight 383.2321
Chemical Name or Material Tandospirone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.