CAS RN 87760-53-0
CAS RN 87760-53-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tandospirone, TRC
CAS: 87760-53-0 Molecular Formula: C21 H29 N5 O2 Molecular Weight (g/mol): 383.49 SMILES: O=C1[C@@H]2[C@H]3CC[C@H](C3)[C@@H]2C(=O)N1CCCCN4CCN(CC4)c5ncccn5
2
Tandospirone, MedChemExpress
MedChemExpress Tandospirone (SM-3997) is a potent and selective 5-HT1A receptor partial agonist, with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.