missing translation for 'onlineSavingsMsg'
Learn More

Tandospirone, MedChemExpress

Product Code. 16446098
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446098 10 mg 10mg
16432237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446098 Supplier Medchem Express Supplier No. HY14558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tandospirone (SM-3997) is a potent and selective 5-HT1A receptor partial agonist, with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.

Tandospirone (SM-3997) is a potent and selective 5-HT1A receptor partial agonist, with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.

Specifications

Chemical Name or Material Tandospirone
CAS 87760-53-0
Color White
Purity Grade Notes Research
Molecular Formula C21H29N5O2
Quantity 10 mg
Synonym SM-3997
Solubility Information DMSO : 16.67 mg/mL (43.47 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.26 mM; Need ultrasonic)
SMILES O=C1[C@]2([H])[C@]([C@H]3CC[C@@H]2C3)([H])C(N1CCCCN(CC4)CCN4C5=NC=CC=N5)=O
Molecular Weight (g/mol) 383.49
Formula Weight 383.49
Percent Purity 96.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.