Learn More
IBA Lifesciences Streptavidin
GREENER_CHOICE
Artikelnummer. 12863186 Alle ansehen IBA Lifesciences Produkte
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
100 mg
1 g
5 g
missing translation for 'unitSize'
100mg
10mg
1g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
12863186 10 mg 10mg
12873186 100 mg 100mg
16491465 1 g 1g
17277730 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12863186 missing translation for 'mfr' IBA Lifesciences missing translation for 'supplierNo' 20203010
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Streptavidin is a widely used tool for immobilization and assays of biotinylated proteins or other biotin conjugated molecules.
Streptavidin is a tetrameric protein composed of identic subunits. Each subunit specifically binds one biotin molecule with fM affinity, which makes it one of the strongest non-covalent interactions known. The preparation contains an N- and C-terminal shortened variant (core streptavidin) with improved properties regarding homogeneity, solubility, resistance towards proteolytic degradation, and accessibility of the biotin binding pocket as compared to native streptavidin.
The streptavidin:biotin system is widely used for immobilization and detection of biotinylated molecules, like proteins and nucleic acids. IBA Lifesciences provides bulk amounts of lyophilized streptavidin in reliable and high quality at competitive prices to generate streptavidin-coated surfaces (microplates, SPR chips, beads) or detection reagents conjugated with, e.g., fluorescent dyes.
Please note that streptavidin is not applicable for detection, immobilization, and purification of Strep-tag™II and Twin-Strep-tag™ fusion proteins, since the binding affinity for both tags is too low.
Spezifikation
Particle Size 13.3 kDa/subunit
Quantity 10 mg
Source Recombinant, expression in E. coli
Purity or Quality Grade >95 %
For Research Use Only.