Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ Resin
Product Code. 16664714 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1 set
1000mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16664714 4 mL 1 set
16674714 20 mL 1 set
16684714 50 mL 1 set
17267810 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16664714 Supplier IBA Lifesciences Supplier No. 25010002
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Strep-Tactin™XT 4Flow™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT 4Flow™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
  • The low concentrated agarose allows for the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
  • In addition, Strep-Tactin™XT 4Flow™ enables the application of denaturing agents such as urea.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary. 
Specifications
Matrix 4% agarose, crosslinked
Particle Size 50 to 150 μm
Quantity 4 mL
Target Twin-Strep-Tag™ and Strep-tag™II
Format Pre-packed Gravity Flow Column
For Research Use Only.