missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
STK33 Polyclonal specifically detects STK33 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Antigen | STK33 |
| Applications | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | Q9BYT3 |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 33, serine/threonine-protein kinase 33 |
| Gene Symbols | STK33 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?