missing translation for 'onlineSavingsMsg'
Learn More

Staphylococcus Aureus Enterotoxin B Antibody (S643), DyLight 550, Novus Biologicals™

Product Code. 18214544 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214544 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214544 Supplier Novus Biologicals Supplier No. NB10073025R
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Staphylococcus Aureus Enterotoxin B Monoclonal specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for ELISA, Sandwich ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus Aureus Enterotoxin B
Applications ELISA
Classification Monoclonal
Clone S643
Conjugate DyLight 550
Dilution ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias S. aureus
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Staphylococcus Aureus Enterotoxin B. This does not cross react with S. aureus enterotoxins A, C, D and E.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.