Learn More
R&D Systems™ Spleen Tissue Slides (Normal)- Paraffin
Product Code. 18759083 Shop All Bio Techne Products
Change view
Click to view available options
Tissue Condition:
Normal
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tissue Condition Unit Size
18759083 Normal Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18759083 Supplier R&D Systems™ Supplier No. NBP230202
  • ONLINE EXCLUSIVE PRICE
    314.00 EUR
    296.10 EUR / Pack of 5
    Save 17.90 EUR
    5% Off
 Restricted Item
Estimated Shipment: 19-11-2026
A Novus product, part of R&D Systems.
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Human
Type Spleen Tissue Slides (Normal)
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Paraffin, In-situ Hybridization, Nucleic Acid Extraction
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Normal
Product Line Novus