Learn More
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device
Product Code. 12035635
Change view
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
20 kDa
3.5 to 5 kDa
50 kDa
8 to 10 kDa
Closure Color:
Black
Blue
Green
Orange
Red
White
Yellow
Height (Metric):
44 mm
63 mm
Unit Size:
Pack of 12
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Closure Color Height (Metric) Unit Size
12035635 100 kDa Blue 63 mm Pack of 12
12005625 0.1 to 0.5 kDa Green 44 mm Pack of 12
12025625 0.5 to 1 kDa Orange 44 mm Pack of 12
12045625 3.5 to 5 kDa Black 44 mm Pack of 12
12065625 8 to 10 kDa Yellow 44 mm Pack of 12
12085625 20 kDa Red 44 mm Pack of 12
12005635 50 kDa White 44 mm Pack of 12
12015625 0.1 to 0.5 kDa Green 63 mm Pack of 12
12025635 100 kDa Blue 44 mm Pack of 12
12035625 0.5 to 1 kDa Orange 63 mm Pack of 12
12055625 3.5 to 5 kDa Black 63 mm Pack of 12
12075625 8 to 10 kDa Yellow 63 mm Pack of 12
12095625 20 kDa Red 63 mm Pack of 12
12015635 50 kDa White 63 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 12035635 Supplier Spectrum™ Supplier No. F235071
  • ONLINE EXCLUSIVE PRICE
    211.00 EUR
    184.00 EUR / Pack of 12
    Save 27.00 EUR
    12% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.
  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume
Specifications
Product Type Micro Float-A-Lyzer Dialysis Device
MWCO 100 kDa
Membrane Cellulose Ester
Material Polycarbonate
Diameter (English) Membrane 0.25 in.
Diameter (Metric) Membrane 6.4 mm
Height (English) 2.4 in.
Height (Metric) 63 mm
Width (English) 1.77 in.
Width (Metric) 45 mm
Capacity (Metric) 400 to 500 μL
Closure Color Blue
Closure Material Polypropylene
Closure Type Luer lok
Kit Contents 1mL syringe
Quantity Each
For Use With (Application) Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis