missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sonic Hedgehog/Shh Polyclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.
Specifications
Specifications
| Antigen | Sonic Hedgehog/Shh |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant mouse Sonic Hedgehog/Shh N-Terminus, Cys25-Gly198 (Lys122Arg), Accession # Q62226 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?