Learn More
Sodium hydrogen fumarate, 98%
Product Code. 11420558
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420558 50 g 50g
11430558 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11420558 Supplier Thermo Scientific Alfa Aesar Supplier No. B24683.18
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
CAS: 5873-57-4 | C4H2NaO4 | 137.05 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5873-57-4
Molecular Formula C4H2NaO4
Molecular Weight (g/mol) 137.05
MDL Number MFCD00039097
InChI Key VRVKOZSIJXBAJG-TYYBGVCCSA-L
Synonym sodium hydrogen fumarate, monosodium fumarate, sodium fumarate, maleic acid, sodium salt, maleinian sodu polish, fumaric acid monosodium salt, ccris 7315, 2-butenedioic acid 2z-, sodium salt, sodium e-3-carboxyacrylate, maleinian sodu
PubChem CID 6076814
IUPAC Name sodium;(E)-4-hydroxy-4-oxobut-2-enoate
SMILES [Na+].[O-]C(=O)\C=C\C([O-])=O
Specifications
Quantity 50 g
Formula Weight 138.06
Percent Purity 98%
Chemical Name or Material Sodium hydrogen fumarate