CAS RN 5873-57-4
IUPAC Name:
sodium (2E)-but-2-enedioate
Synonyms:
sodium fumarate
Molecular Weight (g/mol):
137.05
Molecular Formula:
C4H2NaO4
InChi Key:
VRVKOZSIJXBAJG-TYYBGVCCSA-L
SMILES:
[Na+].[O-]C(=O)\C=C\C([O-])=O
1
–
1
of
1
results
1
Sodium hydrogen fumarate, 98%
CAS: 5873-57-4 Molecular Formula: C4H2NaO4 Molecular Weight (g/mol): 137.05 MDL Number: MFCD00039097 InChI Key: VRVKOZSIJXBAJG-TYYBGVCCSA-L Synonym: sodium hydrogen fumarate,monosodium fumarate,sodium fumarate,maleic acid, sodium salt,maleinian sodu polish,fumaric acid monosodium salt,ccris 7315,2-butenedioic acid 2z-, sodium salt,sodium e-3-carboxyacrylate,maleinian sodu PubChem CID: 6076814 IUPAC Name: sodium;(E)-4-hydroxy-4-oxobut-2-enoate SMILES: [Na+].[O-]C(=O)\C=C\C([O-])=O