Learn More
Sodium Formate, purum p.a, ≥98.0% (NT), Honeywell™
Product Code. 15636800 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15636800 5 kg Poly bottle 5kg
15626800 250 g Poly bottle 250g
15616800 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15636800 Supplier Honeywell Research Chemicals Supplier No. 715405KG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Purum p.a.,≥98.0% (NT)
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
SMILES [Na+].F[B-](F)(F)F
Specifications
Melting Point 255°C
Color Colorless
pH 9.0 to 10.0 (at 20°C)
Physical Form Solid
Quantity 5 kg
Linear Formula NaBF4
UN Number NONH for all modes of transport
Solubility Information 700g/L (at 20°C)
IUPAC Name sodium;tetrafluoroborate
Formula Weight 109.79g/mol
Percent Purity ≥98.0% (T)
Packaging Poly bottle
Density 2.47g/mL at 25°C
Chemical Name or Material Sodium tetrafluoroborate