accessibility menu, dialog, popup

UserName

NaFBFFF

CAS RN 13755-29-8

IUPAC Name: sodium tetrafluoroboranuide
Synonyms: sodium tetrafluoroborate
Molecular Weight (g/mol): 109.79
Molecular Formula: BF4Na
InChi Key: KGJZTOFHXCFQIV-UHFFFAOYSA-N
SMILES: [Na+].F[B-](F)(F)F
Percent Purity 
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)