missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC6A16 Polyclonal specifically detects SLC6A16 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | SLC6A16 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1-4 μg/mL |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gene Alias | NTT5solute carrier family 6 (neurotransmitter transporter), member 16, orphan sodium- and chloride-dependent neurotransmitter transporter NTT5, Solute carrier family 6 member 16, solute carrier family 6, member 16 |
| Gene Symbols | SLC6A16 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SQSFQFPVPWEKCPLTMNSSGFDPECERTTPSIYFWYQQALKASDRIEDGGSP |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?