missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Preparation Method | Use it directly for immuno-precipitation, or heat lysate with SDS gel loading buffer to 95C for 5 minutes followed by rapid cooling for western blot application. If dissociating conditions are required, add reducing agent prior to heating. |
| Quantity | 0.05 mg |
| Content And Storage | Store at -80C. Avoid freeze-thaw cycles. |
| Target | SJCRH30 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?