Learn More
MedChemExpress Sinapine
Click to view available options
:
5 mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Molecular Weight (g/mol) | 310.36 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 18696-26-9 |
| Product Type | Natural Products |
| Purity | 99.0% |
| Solubility | DMSO : 250 mg/mL (805.52 mM∣Need ultrasonic) |
| Target | Cholinesterase (ChE)∣P-glycoprotein |
| Quantity | 5 mg |