Learn More
Tocris Bioscience™ SGC CK2-1
Description
SGC CK2-1 is a potent inhibitor of casein kinase 2 (CK2) with activity at both the CK2A2 and CK2A1 isoforms (IC50 values are 2.3-16 nM and 4.2-36 nM respectively). SGC CK-1 also inhibits DYRK2 (IC50 = 440 nM).
Specifications
Specifications
| Chemical Name or Material | N-[5-[[3-Cyano-7-(cyclopropylamino)pyrazolo[1,5-a]pyrimidin-5-yl]amino]-2-methylphenyl]propanamide |
| CAS | 2470424-39-4 |
| Quantity | 10 mg |
| Target | Casein Kinase 2 Inhibitors |
| Molecular Formula | C20H21N7O |
| Purity | 0.98 |