missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SESN3 Polyclonal specifically detects SESN3 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | SESN3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.50 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000 |
| Formulation | Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide |
| Gene Accession No. | P58005 |
| Gene Alias | SEST3MGC29667, sestrin 3, sestrin-3 |
| Gene Symbols | SESN3 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?