Learn More
Novus Biologicals™ Sambucus Nigra Lectin [Cy5]
Product Code. 18313198 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18313198
missing translation for 'mfr': Novus Biologicals™ CL1305NB
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Concentration 1 mg/mL
For Use With (Application) Immunocytochemistry/Immunofluorescence
Formulation 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM Ca^2+, 0.01 mM Mn^2+, protein stabilizer.
Quantity 1 mg
Storage Requirements Store at 4°C in the dark.
Inhibitor Properties A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37C and centrifuge before use. The recommended concentration range for use is 5-20 μg/mL.
Research Category Glycobiology
Protein Sambucus Nigra Lectin