missing translation for 'onlineSavingsMsg'
Learn More

S-Venlafaxine, TRC

Product Code. 30133171
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133171 50 mg 50mg
30012089 10 mg 10mg
30012416 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133171 Supplier Toronto Research Chemicals Supplier No. TRCV12000850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93413-44-6
Molecular Formula C17 H27 N O2
Molecular Weight (g/mol) 277.4
Synonym Cyclohexanol, 1-[(1S)-2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, Cyclohexanol, 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, (S)-, 1-[(1S)-2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol, (S)-Venlafaxine
IUPAC Name 1-[(1S)-2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol
SMILES COc1ccc(cc1)[C@@H](CN(C)C)C2(O)CCCCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H27NO2/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14/h7-10,16,19H,4-6,11-13H2,1-3H3/t16-/m1/s1
Formula Weight 277.2042
Percent Purity >95%
Chemical Name or Material (S)-Venlafaxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.