CAS RN 93413-44-6
CAS RN 93413-44-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-Venlafaxine, TRC
CAS: 93413-44-6 Molecular Formula: C17 H27 N O2 Molecular Weight (g/mol): 277.4 Synonym: Cyclohexanol, 1-[(1S)-2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-,Cyclohexanol, 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, (S)-,1-[(1S)-2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol,(S)-Venlafaxine IUPAC Name: 1-[(1S)-2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol SMILES: COc1ccc(cc1)[C@@H](CN(C)C)C2(O)CCCCC2